Thermo Scientific Chemicals

Guanylurea sulfate, 98%, Thermo Scientific Chemicals

Catalog number: 411140250
25 g, Plastic bottle, Each
Thermo Scientific Chemicals

Guanylurea sulfate, 98%, Thermo Scientific Chemicals

Catalog number: 411140250
25 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 411140250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
591-01-5
IUPAC Name
bis((diaminomethylidene)urea); sulfuric acid
Molecular Formula
C4H14N8O6S
InChI Key
IATXFPUBPMZBPH-UHFFFAOYSA-N
SMILES
OS(O)(=O)=O.NC(N)=NC(N)=O.NC(N)=NC(N)=O
Appearance (Color)
White
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
96.0 to 104.0 % (On dry substance)
Water
=<10 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text