Hematoxylin, 85%, pure, certified
Hematoxylin, 85%, pure, certified
Hematoxylin, 85%, pure, certified
Hematoxylin, 85%, pure, certified
Thermo Scientific Chemicals

Hematoxylin, 85%, pure, certified

CAS: 517-28-2 | C16H14O6 | 302.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 411161000
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
315.65
Online Exclusive
350.00
Save 34.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS517-28-2
IUPAC Name(1R,10S)-8-oxatetracyclo[8.7.0.0²,⁷.0¹²,¹⁷]heptadeca-2(7),3,5,12(17),13,15-hexaene-5,6,10,14,15-pentol
Molecular FormulaC16H14O6
InChI KeyWZUVPPKBWHMQCE-WKTCHCBJNA-N
SMILES[H][C@]12C3=C(C[C@@]1(O)COC1=C2C=CC(O)=C1O)C=C(O)C(O)=C3
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige to brown
Loss on drying=<10 % (1 g, 105°C, 3 h)
Appearance (Form)Powder
UV spectrophotometry>=80 % (On dry basis)
BSC CertificationPasses
View more
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link