1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals
1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
500 g
Packaging:
View all
Glass bottle
Catalog number 411270050
Price (USD)/ Each
53.65
Online exclusive
59.20 
Save 5.55 (9%)
-
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
53.65
Online exclusive
59.20 
Save 5.55 (9%)
Add to cart
1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals
Catalog number411270050
Price (USD)/ Each
53.65
Online exclusive
59.20 
Save 5.55 (9%)
-
Add to cart
Chemical Identifiers
CAS207300-90-1
IUPAC Namesodium heptane-1-sulfonate hydrate
Molecular FormulaC7H17NaO4S
InChI KeyXWZCREJRXRKIRQ-UHFFFAOYSA-M
SMILESO.[Na+].CCCCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration after Ion exchange>=99.0 %
UVat 210 nm A: =<0.1
UVat 240 nm A: =<0.02
UVat 260 nm A: =<0.01
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only