Thermo Scientific Chemicals

1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals

Catalog number: 411270250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

1-Heptanesulfonic acid sodium salt monohydrate, HPLC grade, Thermo Scientific Chemicals

Catalog number: 411270250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 411270250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
207300-90-1
IUPAC Name
sodium heptane-1-sulfonate hydrate
Molecular Formula
C7H17NaO4S
InChI Key
XWZCREJRXRKIRQ-UHFFFAOYSA-M
SMILES
O.[Na+].CCCCCCCS([O-])(=O)=O
Titration after Ion exchange
>=99.0 %
UV
at 210 nm A: =<0.1
UV
at 240 nm A: =<0.02
UV
at 260 nm A: =<0.01
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text