Thermo Scientific Chemicals

Hexanes, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 411540010
1 L, EcoSafPak™ Glass Bottle, Each
Thermo Scientific Chemicals

Hexanes, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Catalog number: 411540010
1 L, EcoSafPak™ Glass Bottle, Each
Quantity
Packaging
Catalog number: 411540010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
92112-69-1, 110-54-3
Infrared spectrum
Conforms
Appearance (Color)
Clear colorless
Appearance (Form)
Liquid
Refractive index
1.3770 to 1.3810 (20°C, 589 nm)
GC
>=98.5 % (sum of isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text