Thermo Scientific Chemicals

Hexyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 411660250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Hexyl isocyanate, 99%, Thermo Scientific Chemicals

Catalog number: 411660250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 411660250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2525-62-4
IUPAC Name
1-isocyanatohexane
Molecular Formula
C7H13NO
InChI Key
ANJPRQPHZGHVQB-UHFFFAOYSA-N
SMILES
CCCCCCN=C=O
Appearance (Form)
Liquid
Infrared spectrum
Conforms
Appearance (Color)
Colorless to light yellow
Appearance
May develop some turbidity or precipitate
Refractive index
1.4190 to 1.4220 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text