Thermo Scientific Chemicals

Histamine, 97%, Thermo Scientific Chemicals

Catalog number: 411710050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Histamine, 97%, Thermo Scientific Chemicals

Catalog number: 411710050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 411710050
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
51-45-6
IUPAC Name
2-(1H-imidazol-5-yl)ethan-1-amine
Molecular Formula
C5H9N3
InChI Key
NTYJJOPFIAHURM-UHFFFAOYSA-N
SMILES
NCCC1=CN=CN1
Appearance (Form)
Crystalline powder and/or chunks
Infrared spectrum
Conforms
Appearance (Color)
White to light yellow
Melting point
81°C to 85°C
Titration with HClO4
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text