Thermo Scientific Chemicals

DL-Histidine hydrochloride, monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 411720250
25 g, Glass Bottle, Each
Thermo Scientific Chemicals

DL-Histidine hydrochloride, monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 411720250
25 g, Glass Bottle, Each
Quantity
Packaging
Catalog number: 411720250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
123333-71-1
IUPAC Name
hydrogen 2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrate chloride
Molecular Formula
C6H12ClN3O3
InChI Key
CMXXUDSWGMGYLZ-UHFFFAOYNA-N
SMILES
[H+].O.[Cl-].NC(CC1=CN=CN1)C(O)=O
Titration with HClO4
97.5 to 102.5 %
Sulfated ash
=<0.2 %
Solubility
(3 % in H2O) Clear colorless to
Solubility
light yellow solution
Appearance (Color)
White to light yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text