Thermo Scientific Chemicals

L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 411730250
25 g, Plastic Bottle, Each
Thermo Scientific Chemicals

L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: 411730250
25 g, Plastic Bottle, Each
Quantity
Packaging
Catalog number: 411730250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5934-29-2
IUPAC Name
hydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrate chloride
Molecular Formula
C6H12ClN3O3
InChI Key
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILES
[H+].O.[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O
Appearance (Form)
Crystalline powder or crystals
Titration with NaOH
>=97.5 % (On anhydrous substance)
Infrared spectrum
Conforms
Heavy metals (as Pb)
=<10 ppm
Sulfated ash
=<0.1 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text