L-Histidine hydrochloride monohydrate, 98%
L-Histidine hydrochloride monohydrate, 98%
L-Histidine hydrochloride monohydrate, 98%
L-Histidine hydrochloride monohydrate, 98%
Thermo Scientific Chemicals

L-Histidine hydrochloride monohydrate, 98%

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 411731000
Price (USD)
74.65
Special offer
Online exclusive
Ends: 26-Sep-2025
92.40
Save 17.75 (19%)
Each
-
Add to cart
Quantity:
100 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)
74.65
Special offer
Online exclusive
Ends: 26-Sep-2025
92.40
Save 17.75 (19%)
Each
Add to cart
Chemical Identifiers
CAS5934-29-2
IUPAC Name(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
Molecular FormulaC6H12ClN3O3
InChI KeyCMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILESO.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=97.5 % (On anhydrous substance)
Infrared spectrumConforms
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link