L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals
L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number 411735000
Price (USD)/ Each
369.65
Online exclusive
410.00 
Save 40.35 (10%)
-
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
369.65
Online exclusive
410.00 
Save 40.35 (10%)
Add to cart
L-Histidine hydrochloride monohydrate, 98%, Thermo Scientific Chemicals
Catalog number411735000
Price (USD)/ Each
369.65
Online exclusive
410.00 
Save 40.35 (10%)
-
Add to cart
Chemical Identifiers
CAS5934-29-2
IUPAC Namehydrogen (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid hydrate chloride
Molecular FormulaC6H12ClN3O3
InChI KeyCMXXUDSWGMGYLZ-XRIGFGBMSA-N
SMILES[H+].O.[Cl-].N[C@@H](CC1=CN=CN1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Titration with NaOH>=97.5 % (On anhydrous substance)
Infrared spectrumConforms
Heavy metals (as Pb)=<10 ppm
Sulfated ash=<0.1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only