Thermo Scientific Chemicals

Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 411761000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals

Catalog number: 411761000
100 g, Glass bottle, Each
Quantity
Packaging
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 411761000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5341-61-7
IUPAC Name
hydrogen hydrazine chloride
Molecular Formula
ClH5N2
InChI Key
BIVUUOPIAYRCAP-UHFFFAOYSA-N
SMILES
[H+].[Cl-].NN
Appearance (Color)
White
Appearance (Form)
Powder and/or chunks
Infrared spectrum
Conforms
Titration Argentometric
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text