Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals
Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
View all
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 411765000
Price (USD)/ Each
165.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
195.00 
Save 29.35 (15%)
-
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
165.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
195.00 
Save 29.35 (15%)
Add to cart
Hydrazine dihydrochloride, 99%, Thermo Scientific Chemicals
Catalog number411765000
Price (USD)/ Each
165.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
195.00 
Save 29.35 (15%)
-
Add to cart
Chemical Identifiers
CAS5341-61-7
IUPAC Namehydrogen hydrazine chloride
Molecular FormulaClH5N2
InChI KeyBIVUUOPIAYRCAP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NN
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder and/or chunks
Infrared spectrumConforms
Titration Argentometric>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only