Thermo Scientific Chemicals

4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 411781000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydrazinobenzenesulfonic acid, hemihydrate, 98%, Thermo Scientific Chemicals

Catalog number: 411781000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 411781000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
854689-07-9
IUPAC Name
4-hydrazinylbenzene-1-sulfonic acid hydrate
Molecular Formula
C6H10N2O4S
InChI Key
OSOMMFVBTYFMBI-UHFFFAOYSA-N
SMILES
O.NNC1=CC=C(C=C1)S(O)(=O)=O
Appearance (Color)
Light yellow to light orange
Appearance (Form)
Powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %
Loss on drying
4 % to 5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text