Thermo Scientific Chemicals

4-Hydrazinobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 411790250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

4-Hydrazinobenzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: 411790250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 411790250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
619-67-0
IUPAC Name
4-hydrazinylbenzoic acid
Molecular Formula
C7H8N2O2
InChI Key
PCNFLKVWBDNNOW-UHFFFAOYSA-N
SMILES
NNC1=CC=C(C=C1)C(O)=O
Appearance (Color)
Light yellow to light brown
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text