4-Hydrazinobenzoic acid, 98%
4-Hydrazinobenzoic acid, 98%
4-Hydrazinobenzoic acid, 98%
4-Hydrazinobenzoic acid, 98%
Thermo Scientific Chemicals

4-Hydrazinobenzoic acid, 98%

CAS: 619-67-0 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 411791000
Price (USD)
274.00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
274.00
Each
Add to cart
Chemical Identifiers
CAS619-67-0
IUPAC Name4-hydrazinylbenzoic acid
Molecular FormulaC7H8N2O2
InChI KeyPCNFLKVWBDNNOW-UHFFFAOYSA-N
SMILESNNC1=CC=C(C=C1)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to light brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Share catalog number, name or link