Thermo Scientific Chemicals

Acetoin, 96%, may exist as mixture of monomer and dimer, Thermo Scientific Chemicals

Catalog number: 411955000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Acetoin, 96%, may exist as mixture of monomer and dimer, Thermo Scientific Chemicals

Catalog number: 411955000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 411955000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
23147-57-1
IUPAC Name
2,3,5,6-tetramethyl-1,4-dioxane-2,5-diol
Molecular Formula
C8H16O4
InChI Key
DFMGATPNJMFDCR-UHFFFAOYNA-N
SMILES
CC1OC(C)(O)C(C)OC1(C)O
Appearance (Color)
White to yellow
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
GC
>=95 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text