Hydroxylamine hydrochloride, ACS reagent
Hydroxylamine hydrochloride, ACS reagent
Hydroxylamine hydrochloride, ACS reagent
Hydroxylamine hydrochloride, ACS reagent
Thermo Scientific Chemicals

Hydroxylamine hydrochloride, ACS reagent

CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 412051000
Price (USD)
76.65
Online Exclusive
85.40
Save 8.75 (10%)
Each
In stock
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
76.65
Online Exclusive
85.40
Save 8.75 (10%)
Each
Add to cart
Chemical Identifiers
CAS5470-11-1
IUPAC Namehydroxylamine hydrochloride
Molecular FormulaClH4NO
InChI KeyWTDHULULXKLSOZ-UHFFFAOYSA-N
SMILESCl.NO
View more
SpecificationsSpecification SheetSpecification Sheet
Ammonium (NH4)Passes test
Appearance (Color)White
Appearance (Form)Crystalline powder or crystals and/or chunks
Infrared spectrumConforms
Titration with KMnO4>=96.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link