Thermo Scientific Chemicals

(4-Hydroxyphenoxy)acetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 412090250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

(4-Hydroxyphenoxy)acetic acid, 98%, Thermo Scientific Chemicals

Catalog number: 412090250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 412090250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1878-84-8
IUPAC Name
2-(4-hydroxyphenoxy)acetate
Molecular Formula
C8H7O4
InChI Key
PKGWLCZTTHWKIZ-UHFFFAOYSA-M
SMILES
OC1=CC=C(OCC([O-])=O)C=C1
Appearance (Color)
Off-white to beige
Appearance (Form)
Crystalline powder
Infrared spectrum
Conforms
Melting point
152°C to 156°C
Titration with NaOH
>=97.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text