Thermo Scientific Chemicals

(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 412120010
1 g, Glass bottle, Each
Thermo Scientific Chemicals

(S)-3-Hydroxy-3-phenylpropanoic acid, 99%, Thermo Scientific Chemicals

Catalog number: 412120010
1 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 412120010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
36567-72-3
IUPAC Name
(3S)-3-hydroxy-3-phenylpropanoate
Molecular Formula
C9H9O3
InChI Key
AYOLELPCNDVZKZ-QMMMGPOBSA-M
SMILES
O[C@@H](CC([O-])=O)C1=CC=CC=C1
Appearance (Color)
White to off-white
Appearance (Form)
Crystals or needles
Infrared spectrum
Conforms
Melting point
116°C to 119°C
Titration with NaOH
>=98.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text