4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals
4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Hydroxyphthalic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 412130250
Price (USD)/ Each
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-hydroxybenzene-1,2-dicarboxylic acid
Molecular FormulaC8H6O5
InChI KeyMWRVRCAFWBBXTL-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(O)C=C1C(O)=O
Molecular Weight (g/mol)182.13
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only