Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog number 412150010
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
Add to cart
Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals
Catalog number412150010
Price (USD)/ Each
90.65
Online exclusive
101.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS27813-02-1
IUPAC Name1-hydroxypropan-2-yl 2-methylprop-2-enoate
Molecular FormulaC7H12O3
InChI KeyZMARGGQEAJXRFP-UHFFFAOYNA-N
SMILESCC(CO)OC(=O)C(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<20 APHA
Infrared spectrumConforms
Refractive index1.4460 to 1.4490 (20°C, 589 nm)
GC>=97.0 % (Mixture of isomers)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only