Thermo Scientific Chemicals

Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals

Catalog number: 412152500
250 g, Glass bottle, Each
Thermo Scientific Chemicals

Hydroxypropyl methacrylate, 97+%, mixture of isomers, stabilized, Thermo Scientific Chemicals

Catalog number: 412152500
250 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 412152500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
27813-02-1
IUPAC Name
1-hydroxypropan-2-yl 2-methylprop-2-enoate
Molecular Formula
C7H12O3
InChI Key
ZMARGGQEAJXRFP-UHFFFAOYNA-N
SMILES
CC(CO)OC(=O)C(C)=C
Appearance (Form)
Clear liquid
Color scale
=<20 APHA
Infrared spectrum
Conforms
Refractive index
1.4460 to 1.4490 (20°C, 589 nm)
GC
>=97.0 % (Mixture of isomers)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text