Chemical Identifiers
CAS6358-69-6
IUPAC Nametrisodium 8-hydroxypyrene-1,3,6-trisulfonate
Molecular FormulaC16H7Na3O10S3
InChI KeyKXXXUIKPSVVSAW-UHFFFAOYSA-K
SMILES[Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
View more