Thermo Scientific Chemicals

8-Hydroxy-1,3,6-pyrenetrisulfonic acid trisodium salt, 98%

Catalog number: 412180250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

8-Hydroxy-1,3,6-pyrenetrisulfonic acid trisodium salt, 98%

Catalog number: 412180250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 412180250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6358-69-6
IUPAC Name
trisodium 8-hydroxypyrene-1,3,6-trisulfonate
Molecular Formula
C16H7Na3O10S3
InChI Key
KXXXUIKPSVVSAW-UHFFFAOYSA-K
SMILES
[Na+].[Na+].[Na+].OC1=CC(=C2C=CC3=C(C=C(C4=CC=C1C2=C34)S([O-])(=O)=O)S([O-])(=O)=O)S([O-])(=O)=O
Appearance (Form)
Powder or crystals
Infrared spectrum
Conforms
UV Lambda max
450 to 458 nm (in 0.01N NaOH)
UV spectrophotometry
>=97.5 %
Appearance (Color)
Yellow to yellow-green

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text