Thermo Scientific Chemicals

Iodine monochloride, ACS reagent, Thermo Scientific Chemicals

Catalog number: 412361000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Iodine monochloride, ACS reagent, Thermo Scientific Chemicals

Catalog number: 412361000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 412361000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7790-99-0
IUPAC Name
iodochlorane
Molecular Formula
ClI
InChI Key
QZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILES
ClI
Appearance (Color)
Dark red-brown to brown to black
Assay
1.00 to 1.20 (I/Cl ratio of the Wijs solution
Insoluble matter
=<0.005 %
Assay
prepared from the reagent)
Appearance (Form)
Liquid or solid

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text