4-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals
4-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Iodobenzoyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 412390050
Price (USD)/ Each
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Name4-iodobenzoyl chloride
Molecular FormulaC7H4ClIO
InChI KeyNJAKCIUOTIPYED-UHFFFAOYSA-N
SMILESClC(=O)C1=CC=C(I)C=C1
Molecular Weight (g/mol)266.46
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to light brown
Appearance (Form)Crystals or powder or crystalline powder
Appearance (Form)and/or chunks
Infrared spectrumConforms
Melting point61°C to 68°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only