Chemical Identifiers
CAS3599-32-4
IUPAC Namesodium 2-[(5E)-7-[(2E)-1,1-dimethyl-3-(4-sulfonatobutyl)-1H,2H,3H-benzo[e]indol-2-ylidene]hepta-1,3,5-trien-1-yl]-1,1-dimethyl-3-(4-sulfonatobutyl)-1H-benzo[e]indol-3-ium
Molecular FormulaC43H47N2NaO6S2
InChI KeyMOFVSTNWEDAEEK-UHFFFAOYSA-M
SMILES[Na+].CC1(C)C(=C/C=C/C=CC=CC2=[N+](CCCCS([O-])(=O)=O)C3=CC=C4C=CC=CC4=C3C2(C)C)N(CCCCS([O-])(=O)=O)C2=CC=C3C=CC=CC3=C12
View more