Thermo Scientific Chemicals

Lanthanum(III) oxide, 91%, Thermo Scientific Chemicals

Catalog number: 413022500
250 g, Each
Thermo Scientific Chemicals

Lanthanum(III) oxide, 91%, Thermo Scientific Chemicals

Catalog number: 413022500
250 g, Each
Quantity
Catalog number: 413022500
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1312-81-8
IUPAC Name
dilanthanum(3+) trioxidandiide
Molecular Formula
La2O3
InChI Key
MRELNEQAGSRDBK-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[La+3].[La+3]
Appearance (Form)
Powder
Appearance (Color)
White to light grey
Assay
>=89 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text