Lissamine Rhodamine B Sulfonyl Chloride, +99%
Lissamine Rhodamine B Sulfonyl Chloride, +99%
Thermo Scientific Chemicals

Lissamine Rhodamine B Sulfonyl Chloride, +99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 413230010
Price (USD)/ Each
355.65
Online exclusive
395.00 
Save 39.35 (10%)
-
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
355.65
Online exclusive
395.00 
Save 39.35 (10%)
Add to cart
Lissamine Rhodamine B Sulfonyl Chloride, +99%
Catalog number413230010
Price (USD)/ Each
355.65
Online exclusive
395.00 
Save 39.35 (10%)
-
Add to cart
Chemical Identifiers
CAS62796-29-6
IUPAC Name9-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium
Molecular FormulaC27H29ClN2O6S2
InChI KeyYERWMQJEYUIJBO-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S(Cl)(=O)=O)=C2C=C1)N(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Molar extinction coefficient>=85000 l/(mol.cm) (lambda max)
Appearance (Color)Dark green
Appearance (Form)Crystalline powder
UV Lambda max561 to 567 nm (in CH2Cl2)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only