Chemical Identifiers
CAS62796-29-6
IUPAC Name9-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium
Molecular FormulaC27H29ClN2O6S2
InChI KeyYERWMQJEYUIJBO-UHFFFAOYSA-N
SMILESCCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S(Cl)(=O)=O)=C2C=C1)N(CC)CC
View more