Thermo Scientific Chemicals

Lithium dodecyl sulfate, +99%

Catalog number: 413300250
25 g, Glass bottle, Each
Thermo Scientific Chemicals

Lithium dodecyl sulfate, +99%

Catalog number: 413300250
25 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 413300250
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2044-56-6
IUPAC Name
lithium(1+) dodecyl sulfate
Molecular Formula
C12H25LiO4S
InChI Key
YFVGRULMIQXYNE-UHFFFAOYSA-M
SMILES
[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
Infrared spectrum
Conforms
UV
(5 % in water) 1 cm cell
Appearance (Color)
White
Titration after Ion exchange
>=99.0 %
UV
at 260 nm A: =<0.1

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text