Lithium dodecyl sulfate, +99%
Lithium dodecyl sulfate, +99%
Thermo Scientific Chemicals

Lithium dodecyl sulfate, +99%

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number 413301000
Price (USD)/ Each
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
IUPAC Namelithium(1+) dodecyl sulfate
Molecular FormulaC12H25LiO4S
InChI KeyYFVGRULMIQXYNE-UHFFFAOYSA-M
SMILES[Li+].CCCCCCCCCCCCOS([O-])(=O)=O
Molecular Weight (g/mol)272.33
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
UV(5 % in water) 1 cm cell
Appearance (Color)White
Titration after Ion exchange>=99.0 %
UVat 260 nm A: =<0.1
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only