Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
100 g
500 g
Packaging:
Glass bottle
Catalog number 413320025
Price (USD)/ Each
821.65
Online exclusive
913.00 
Save 91.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)/ Each
821.65
Online exclusive
913.00 
Save 91.35 (10%)
Add to cart
Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
Catalog number413320025
Price (USD)/ Each
821.65
Online exclusive
913.00 
Save 91.35 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Titration with HCl>=98.0 %
Heavy metals (ICP-OES)=<0.002 %
Sulfate (SO4)=<0.05 %
Lithium carbonate=<2.0 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only