Thermo Scientific Chemicals

Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 413325000
500 g, Glass bottle, Each
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Catalog number: 413325000
500 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 413325000
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1310-66-3
IUPAC Name
lithium(1+) hydrate hydroxide
Molecular Formula
H3LiO2
InChI Key
GLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES
[Li+].O.[OH-]
Appearance (Color)
White
Titration with HCl
>=98.0 %
Heavy metals (ICP-OES)
=<0.002 %
Sulfate (SO4)
=<0.05 %
Lithium carbonate
=<2.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text