Thermo Scientific Chemicals

Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals

Catalog number: 413330050
5 g, Glass bottle, Each
Thermo Scientific Chemicals

Lactic acid, lithium salt, 99%, Thermo Scientific Chemicals

Catalog number: 413330050
5 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 413330050
Price (USD)
Have Questions?

Chemical Identifiers

CAS
867-55-0
IUPAC Name
lithium(1+) 2-hydroxypropanoate
Molecular Formula
C3H5LiO3
InChI Key
GKQWYZBANWAFMQ-UHFFFAOYNA-M
SMILES
[Li+].CC(O)C([O-])=O
Appearance (Color)
White
Titration with HClO4
98.5 to 101.5 %
Appearance (Form)
Crystalline powder
pH
6 to 9 (1% solution)
Solubility
(10% in water) Clear colorless

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text