Thermo Scientific Chemicals

Lithium stearate, 95%, Thermo Scientific Chemicals

Catalog number: 413341000
100 g, Glass bottle, Each
Thermo Scientific Chemicals

Lithium stearate, 95%, Thermo Scientific Chemicals

Catalog number: 413341000
100 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 413341000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
4485-12-5
IUPAC Name
lithium(1+) octadecanoate
Molecular Formula
C18H35LiO2
InChI Key
HGPXWXLYXNVULB-UHFFFAOYSA-M
SMILES
[Li+].CCCCCCCCCCCCCCCCCC([O-])=O
Appearance (Color)
White
Titration with HClO4
>=94 %
Appearance (Form)
Powder
Infrared spectrum
Conforms

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text