Thermo Scientific Chemicals

DL-Lysine Monohydrochloride 99%, Thermo Scientific Chemicals

Catalog number: 413370010
1 kg, Glass Bottle, Each
Thermo Scientific Chemicals

DL-Lysine Monohydrochloride 99%, Thermo Scientific Chemicals

Catalog number: 413370010
1 kg, Glass Bottle, Each
Quantity
Packaging
Catalog number: 413370010
Price (USD)
Have Questions?

Chemical Identifiers

CAS
70-53-1
IUPAC Name
2,6-diaminohexanoic acid hydrochloride
Molecular Formula
C6H15ClN2O2
InChI Key
BVHLGVCQOALMSV-UHFFFAOYSA-N
SMILES
Cl.NCCCCC(N)C(O)=O
Infrared spectrum
Conforms
Specific optical rotation
-1° to +1° (20°C, 589 nm) (c=1, water)
Appearance (Form)
Powder
Appearance (Color)
White to off-white
Titration with HClO4
98.5 to 101.5 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text