Thermo Scientific Chemicals

Magnesium carbonate basic, heavy, extra pure, Thermo Scientific Chemicals

Catalog number: 413400025
2.5 kg, Plastic Drum, Each
Thermo Scientific Chemicals

Magnesium carbonate basic, heavy, extra pure, Thermo Scientific Chemicals

Catalog number: 413400025
2.5 kg, Plastic Drum, Each
Quantity
Packaging
Catalog number: 413400025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
39409-82-0
IUPAC Name
pentamagnesium(2+) tetrahydrate dihydroxide tetracarbonate
Molecular Formula
C4H10Mg5O18
InChI Key
YMKSTRLFWJQKFP-UHFFFAOYSA-D
SMILES
O.O.O.O.[OH-].[OH-].[Mg++].[Mg++].[Mg++].[Mg++].[Mg++].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O
Appearance (Color)
White to almost white
Appearance (Form)
Powder
Titration Complexometric
40 to 45 % (MgO)
Soluble salts
=<1.0 %
Calcium oxide (CaO)
=<0.6 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text