Thermo Scientific Chemicals

Magnesium perchlorate, ACS reagent, desiccant, Thermo Scientific Chemicals

Catalog number: 413465000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Magnesium perchlorate, ACS reagent, desiccant, Thermo Scientific Chemicals

Catalog number: 413465000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 413465000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
10034-81-8
IUPAC Name
magnesium(2+) diperchlorate
Molecular Formula
Cl2MgO8
InChI Key
MPCRDALPQLDDFX-UHFFFAOYSA-L
SMILES
[Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Appearance (Form)
Crystalline powder or flakes and/or chunks
Titrable free acid
=<0.005 meq/g
Suitability
(for moisture absorption) Passes test
Appearance (Color)
White
Loss on drying
=<8 % (1 g, 190°C)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent, Oxidation reagent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text