Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals
Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals

Magnesium sulfate, 97%, MgO4S, CAS Number-7487-88-9, sulfuric acid magnesium salt 1:1, sal de sedlitz, magnesium sulfate, tomix ot, bitter salt, magnesium sulfate 1:1, sal angalis, magnesium sulphate, mgso4, magnesium sulfate anhydrous, 500g, CHEBI:32599, Undesignated, >700 deg.C
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 kg
5 g
500 g
25 kg
Packaging:
View all
Plastic Bottle
Catalog number 413480025
Price (USD)/ Each
270.65
Online exclusive
301.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
2.5 kg
Packaging:
Plastic Bottle
Request bulk or custom format
Price (USD)/ Each
270.65
Online exclusive
301.00 
Save 30.35 (10%)
Add to cart
Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals
Catalog number413480025
Price (USD)/ Each
270.65
Online exclusive
301.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7487-88-9
IUPAC Namemagnesium(2+) sulfate
Molecular FormulaMgO4S
InChI KeyCSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain grey particles
Appearance (Color)White to light grey
Appearance (Form)Granular powder
Titration Complexometric>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only