1x1 image pixel for data collection
Magnesium sulfate, 97%, pure, anhydrous
Magnesium sulfate, 97%, pure, anhydrous
Magnesium sulfate, 97%, pure, anhydrous
Magnesium sulfate, 97%, pure, anhydrous
Thermo Scientific Chemicals

Magnesium sulfate, 97%, pure, anhydrous

Magnesium sulfate, 97%, MgO4S, CAS Number-7487-88-9, sulfuric acid magnesium salt 1:1, sal de sedlitz, magnesium sulfate, tomix ot, bitter salt, magnesium sulfate 1:1, sal angalis, magnesium sulphate, mgso4, magnesium sulfate anhydrous, 500g, CHEBI:32599, Undesignated, >700 deg.C | CAS: 7487-88-9 | MgO4S | 120.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
500 g
2.5 kg
25 kg
Packaging:
View all
Glass Bottle
Catalog number 413480050
Price (USD)
44.65
Online Exclusive
49.60
Save 4.95 (10%)
Each
-
Add to cart
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
44.65
Online Exclusive
49.60
Save 4.95 (10%)
Each
Add to cart
Chemical Identifiers
CAS7487-88-9
IUPAC Namemagnesium(2+) sulfate
Molecular FormulaMgO4S
InChI KeyCSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES[Mg++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
AppearanceMay contain grey particles
Appearance (Color)White to light grey
Appearance (Form)Granular powder
Titration Complexometric>=96.0 %
Ask our AI about this Product
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
A0473036Certificate of AnalysisJun 11, 2025413480050, 413485000, 413480250, 413480025
B0157010Certificate of AnalysisJun 09, 2025413480050, 413485000, 413480250, 413480025
A0445609Certificate of AnalysisMay 17, 2025413480050, 413485000, 413480250, 413480025
A0428993Certificate of AnalysisMay 17, 2025413480050, 413485000, 413480250, 413480025
A0468734Certificate of AnalysisJan 31, 2025413480050, 413480250, 413485000, 413480025
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection