Thermo Scientific Chemicals

Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 413480250
25 kg, Fibre drum, Each
Thermo Scientific Chemicals

Magnesium sulfate, 97%, pure, anhydrous, Thermo Scientific Chemicals

Catalog number: 413480250
25 kg, Fibre drum, Each
Quantity
Packaging
Catalog number: 413480250
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7487-88-9
IUPAC Name
magnesium(2+) sulfate
Molecular Formula
MgO4S
InChI Key
CSNNHWWHGAXBCP-UHFFFAOYSA-L
SMILES
[Mg++].[O-]S([O-])(=O)=O
Appearance
May contain grey particles
Appearance (Color)
White to light grey
Appearance (Form)
Granular powder
Titration Complexometric
>=96.0 %

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Adsorbent
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text