1x1 image pixel for data collection
Malachite Green oxalate, pure, certified
Malachite Green oxalate, pure, certified
Malachite Green oxalate, pure, certified
Malachite Green oxalate, pure, certified
Thermo Scientific Chemicals

Malachite Green oxalate, pure, certified

CAS: 2437-29-8 | C52H54N4O12 | 927.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number 413490250
Price (USD)
46.65
Online Exclusive
51.70
Save 5.05 (10%)
Each
Add to cart
Quantity:
25 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
46.65
Online Exclusive
51.70
Save 5.05 (10%)
Each
Add to cart
Chemical Identifiers
CAS2437-29-8
IUPAC Namebis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
Molecular FormulaC52H54N4O12
InChI KeyCNYGFPPAGUCRIC-UHFFFAOYSA-L
SMILESOC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Visual transition intervalfrom pH 0.0 (yellow) to pH 2.0 (green)
Appearance (Color)Green to dark green
Infrared spectrumConforms
UV Lambda max614 to 620 nm (in water)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link