Chemical Identifiers
CAS2437-29-8
IUPAC Namebis(bis[4-(dimethylamino)phenyl](phenyl)methylium) oxalic acid dihydrogen oxalate
Molecular FormulaC52H54N4O12
InChI KeyCNYGFPPAGUCRIC-UHFFFAOYSA-L
SMILESOC(=O)C(O)=O.OC(=O)C([O-])=O.OC(=O)C([O-])=O.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C.CN(C)C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=C(C=C1)N(C)C
View more