Thermo Scientific Chemicals

Malonic acid disodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 413505000
500 g, Plastic bottle, Each
Thermo Scientific Chemicals

Malonic acid disodium salt hydrate, 99%, Thermo Scientific Chemicals

Catalog number: 413505000
500 g, Plastic bottle, Each
Quantity
Packaging
Catalog number: 413505000
Price (USD)
Have Questions?

Chemical Identifiers

CAS
132499-82-2
IUPAC Name
disodium propanedioate
Molecular Formula
C3H2Na2O4
InChI Key
PRWXGRGLHYDWPS-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)CC([O-])=O
Appearance (Color)
White
Titration with HClO4
>=98.5 % (On dry substance)
Appearance (Form)
Crystalline powder and/or chunks
Infrared spectrum
Conforms
Water
7 to 13 % (K.F.)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text