Thermo Scientific Chemicals

Menthofuran, 95%, Thermo Scientific Chemicals

Catalog number: 413560100
10 g, Glass bottle, Each
Thermo Scientific Chemicals

Menthofuran, 95%, Thermo Scientific Chemicals

Catalog number: 413560100
10 g, Glass bottle, Each
Quantity
Packaging
Catalog number: 413560100
Price (USD)
Have Questions?

Chemical Identifiers

CAS
494-90-6
IUPAC Name
(6R)-3,6-dimethyl-4,5,6,7-tetrahydro-1-benzofuran
Molecular Formula
C10H14O
InChI Key
YGWKXXYGDYYFJU-SSDOTTSWSA-N
SMILES
C[C@@H]1CCC2=C(C1)OC=C2C
Appearance (Form)
Liquid
Appearance (Color)
Clear colorless to yellow
GC
>=94 %
Infrared spectrum
Conforms
Refractive index
1.4810 to 1.4870 (20°C, 589 nm)

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text