Thermo Scientific Chemicals

Mercury(II) sulfate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 413650025
2.5 kg, Plastic bottle, Each
Thermo Scientific Chemicals

Mercury(II) sulfate, ACS reagent, Thermo Scientific Chemicals

Catalog number: 413650025
2.5 kg, Plastic bottle, Each
Quantity
Packaging
Catalog number: 413650025
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7783-35-9
IUPAC Name
mercury(2+) sulfate
Molecular Formula
HgO4S
InChI Key
DOBUSJIVSSJEDA-UHFFFAOYSA-L
SMILES
[Hg++].[O-]S([O-])(=O)=O
Appearance (Color)
White to off-white
Titration with NH4SCN
>=98.0 %
Nitrate (NO3)
Passes test (limit about 0.003%)
Chloride (Cl)
=<30 ppm
Iron (Fe)
=<50 ppm

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text