Metanil Yellow
Thermo Scientific Chemicals

Metanil Yellow

 
Have Questions?
Quantity:
1 kg
Packaging:
Glass bottle
Catalog number 413720010
Price (USD)/ Each
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS587-98-4
IUPAC Namesodium 3-{2-[4-(phenylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular FormulaC18H14N3NaO3S
InChI KeyNYGZLYXAPMMJTE-UHFFFAOYSA-M
SMILES[Na+].[O-]S(=O)(=O)C1=CC=CC(=C1)N=NC1=CC=C(NC2=CC=CC=C2)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV Lambda max413 to 419 nm (in methanol)
Appearance (Color)Golden to orange
Visual transition intervalfrom pH 1.2 (red) to 2.3 (yellow)
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only