Methanol, ≥99.8%, for spectroscopy ACS
Methanol, ≥99.8%, for spectroscopy ACS
Methanol, ≥99.8%, for spectroscopy ACS
Methanol, ≥99.8%, for spectroscopy ACS
Thermo Scientific Chemicals

Methanol, ≥99.8%, for spectroscopy ACS

CAS: 67-56-1 | CH4O | 32.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number 413775000
Price (USD)
41.65
Online Exclusive
45.90
Save 4.25 (9%)
Each
-
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (USD)
41.65
Online Exclusive
45.90
Save 4.25 (9%)
Each
Add to cart
Chemical Identifiers
CAS67-56-1
IUPAC Namemethanol
Molecular FormulaCH4O
InChI KeyOKKJLVBELUTLKV-UHFFFAOYSA-N
SMILESCO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Carbonyl compounds=<0.001 %
Titrable acid=<0.0003 meq/g
Titrable base=<0.0002 meq/g
KMnO4-reducing substancePasses test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For UV/VIS spectroscopy
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection